Bis(pentafluorophenyl) carbonate, 98+%
Bis(pentafluorophenyl) carbonate, 98+%
Bis(pentafluorophenyl) carbonate, 98+%
Thermo Scientific Chemicals

Bis(pentafluorophenyl) carbonate, 98+%

CAS: 59483-84-0 | C13F10O3 | 394.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H54880.06
also known as H54880-06
Price (EUR)
168,65
Online exclusive
187,00
Save 18,35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
168,65
Online exclusive
187,00
Save 18,35 (10%)
Each
Chemical Identifiers
CAS59483-84-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>98.0%
Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.

Solubility
Soluble in chloroform, ethyl acetate and methylene chloride.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Grošelj, U.; Pušavec, E.; Golobič, A.; Dahmann, G.; Stanovnik, B.; Svete, J. Synthesis of 1,5-disubstituted-4-oxo-4,5-dihydro-1H-pyrazolo[4,3-c]pyridine-7-carboxamides. Tetrahedron 2015, 71 (1), 109-123.
  2. Olsén, P.; Odelius, K.; Albertsson, A. C. Ring-Closing Depolymerization: A Powerful Tool for Synthesizing the Allyloxy-Functionalized Six-Membered Aliphatic Carbonate Monomer 2-Allyloxymethyl-2-ethyltrimethylene Carbonate. Macromolecules 2014, 47 (18), 6189-6195.