3-Bromo-5-fluorophenol, 97%
3-Bromo-5-fluorophenol, 97%
3-Bromo-5-fluorophenol, 97%
3-Bromo-5-fluorophenol, 97%
Thermo Scientific Chemicals

3-Bromo-5-fluorophenol, 97%

CAS: 433939-27-6 | C6H4BrFO | 190.999 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55288.03
also known as H55288-03
Price (EUR)
20,70
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
20,70
Each
Add to cart
Chemical Identifiers
CAS433939-27-6
IUPAC Name3-bromo-5-fluorophenol
Molecular FormulaC6H4BrFO
InChI KeyJCPJGUPQZDEZQH-UHFFFAOYSA-N
SMILESOC1=CC(F)=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only