4-Amino-3-bromobenzonitrile, 97%
4-Amino-3-bromobenzonitrile, 97%
4-Amino-3-bromobenzonitrile, 97%
Thermo Scientific Chemicals

4-Amino-3-bromobenzonitrile, 97%

CAS: 50397-74-5 | C7H5BrN2 | 197.035 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55483.03
also known as H55483-03
Price (EUR)
46,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
46,70
Each
Chemical Identifiers
CAS50397-74-5
IUPAC Name4-amino-3-bromobenzonitrile
Molecular FormulaC7H5BrN2
InChI KeyPOESQIHWIIWNJL-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1Br)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream
Assay from Supplier's CofA≥96.0%
Mass SpecConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only