4-Chlorobenzeneboronic acid pinacol ester, 97%
4-Chlorobenzeneboronic acid pinacol ester, 97%
4-Chlorobenzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

4-Chlorobenzeneboronic acid pinacol ester, 97%

CAS: 195062-61-4 | C12H16BClO2 | 238.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H55984.03
also known as H55984-03
Price (EUR)
26,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
26,30
Each
Specifications
CAS195062-61-4
Chemical Name or Material4-Chlorobenzeneboronic acid pinacol ester
ColorBrown to White
Flash Point>110°C (230°F)
Formula Weight238.52
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only