3-Boc-3-(methylamino)azetidine hydrochloride, 95%
3-Boc-3-(methylamino)azetidine hydrochloride, 95%
3-Boc-3-(methylamino)azetidine hydrochloride, 95%
Thermo Scientific Chemicals

3-Boc-3-(methylamino)azetidine hydrochloride, 95%

CAS: 577777-20-9 | C9H18N2O2 | 186.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H56060.03
also known as H56060-03
Price (EUR)
234,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
234,00
Each
Chemical Identifiers
CAS577777-20-9
SpecificationsSpecification SheetSpecification Sheet
Melting Point167-171?C
Proton NMRConforms
FormPowder
Appearance (Color)White to cream
Assay from Supplier's CofA≥94.0%
It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water. (5 g/100ml)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
RUO – Research Use Only
Mounir Andalouss.; Herman D. Lim.; Tiffany van der Meer.; Maarten Sijm.; Chris B.M. Poulie.; Iwan J.P. de Esch.; Rob Leurs.; Rogier A. Smits. A novel series of histamine H4 receptor antagonists based on the pyrido[3,2-d]pyrimidine scaffold: Comparison of hERG binding and target residence time with PF-3893787.Bioorg. Med. Chem. Lett. 2013, 23 (9),2663-2670 .