2-Fluoro-N-methylbenzamide, 97%
2-Fluoro-N-methylbenzamide, 97%
2-Fluoro-N-methylbenzamide, 97%
Thermo Scientific Chemicals

2-Fluoro-N-methylbenzamide, 97%

CAS: 52833-63-3 | C8H8FNO | 153.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H56104.03
also known as H56104-03
Price (EUR)
260,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
260,00
Each
Chemical Identifiers
CAS52833-63-3
IUPAC Name2-fluoro-N-methylbenzamide
Molecular FormulaC8H8FNO
InChI KeyNAGFMACWWJYORB-UHFFFAOYSA-N
SMILESCNC(=O)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only