3,4-Dihydro-2H-pyran-2-methanol, 96%
3,4-Dihydro-2H-pyran-2-methanol, 96%
3,4-Dihydro-2H-pyran-2-methanol, 96%
Thermo Scientific Chemicals

3,4-Dihydro-2H-pyran-2-methanol, 96%

CAS: 3749-36-8 | C6H10O2 | 114.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H56124.03
also known as H56124-03
Price (EUR)
39,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
39,50
Each
Specifications
CAS3749-36-8
Chemical Name or Material3,4-Dihydro-2H-pyran-2-methanol
Density1.101
EINECS Number223-150-0
Flash Point89°C (192°F)
View more
It is used in the preparation of supported dihydropyran for mild immobilization and cleavage of alcohols

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of supported dihydropyran for mild immobilization and cleavage of alcohols

Notes
Incompatible with oxidizing agents, acids and heats. Store at room temperature.
RUO – Research Use Only

General References:

  1. Curtis W. Smith.; Douglas G. Norton.; Seaver A. Ballard. Reactions of Acrolein and Related Compounds. II. Addition of Unsaturated Esters and of Methacrylonitrile. J. Am. Chem. Soc. 1951, 73 (11),5270-5272.
  2. F. Sweet.; R. K. Brown. Synthesis of 1,6-anhydro-4-deoxy-β-DL-xylo-hexopyranose. Can. J. Chem. 1968, 46 (13),2289-2298.