Methyl 2-oxoindane-1-carboxylate, 97%
Methyl 2-oxoindane-1-carboxylate, 97%
Methyl 2-oxoindane-1-carboxylate, 97%
Thermo Scientific Chemicals

Methyl 2-oxoindane-1-carboxylate, 97%

CAS: 104620-34-0 | C11H10O3 | 190.198 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H57535.03
also known as H57535-03
Price (EUR)
62,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
62,10
Each
Chemical Identifiers
CAS104620-34-0
IUPAC Namemethyl 2-oxo-2,3-dihydro-1H-indene-1-carboxylate
Molecular FormulaC11H10O3
InChI KeyRWSYHHRDYRDVQL-UHFFFAOYNA-N
SMILESCOC(=O)C1C(=O)CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow or pale brown to brown
FormPowder or crystals
Assay (HPLC)≥96.0%
Water Content (Karl Fischer Titration)<0.5%
Melting point62-75°C
View more
It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
RUO – Research Use Only