n-Butyl levulinate, 98%
n-Butyl levulinate, 98%
n-Butyl levulinate, 98%
Thermo Scientific Chemicals

n-Butyl levulinate, 98%

CAS: 2052-15-5 | C9H16O3 | 172.224 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number H59274.AE
also known as H59274-AE
Price (EUR)
22,40
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
22,40
Each
Chemical Identifiers
CAS2052-15-5
IUPAC Namebutyl 4-oxopentanoate
Molecular FormulaC9H16O3
InChI KeyISBWNEKJSSLXOD-UHFFFAOYSA-N
SMILESCCCCOC(=O)CCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
Butyl levulinate was used in the synthesis of γ-valerolactone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyl levulinate was used in the synthesis of γ-valerolactone.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Xian-Long Du; Qing-Yuan Bi; Yong-Mei Liu; Yong Cao; Kang-Nian Fan. Conversion of biomass-derived levulinate and formate esters into γ-valerolactone over supported gold catalysts. Chemsuschem Chemistry And Sustainability, Energy & Materials,2011,4(12), 1838-1843.
  2. S. Dharne; V.V. Bokade. Esterification of levulinic acid to n-butyl levulinate over heteropolyacid supported on acid-treated clay. Journal of Natural Gas Chemistry,2011,20(1), 18-24.