1-Ethyl-3-methylimidazolium acetate, 97%
1-Ethyl-3-methylimidazolium acetate, 97%
1-Ethyl-3-methylimidazolium acetate, 97%
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium acetate, 97%

CAS: 143314-17-4 | C8H14N2O2 | 170.212 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H59277.18
also known as H59277-18
Price (EUR)
853,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
853,00
Each
Chemical Identifiers
CAS143314-17-4
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium acetate
Molecular FormulaC8H14N2O2
InChI KeyXIYUIMLQTKODPS-UHFFFAOYSA-M
SMILESCC([O-])=O.CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to orange to brown
Assay from Supplier's CofA≥96.0% (HPLC)
Assay from Supplier's CofA≥96.0% (HPLC)
FormLiquid
Appearance (Color)Pale yellow to orange to brown
View more
It is used as a solvent and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a solvent and as pharmaceutical intermediate.

Solubility
Soluble in water, acetonitrile, acetone, dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Ning Sun.; Mustafizur Rahman.; Ying Qin.; Mirela L. Maxim.; Héctor Rodríguez.; Robin D. Rogers. Complete dissolution and partial delignification of wood in the ionic liquid 1-ethyl-3-methylimidazolium acetate.Green Chem. 2009, 11 (5),646-655.
  2. Jing-Fang Huang.; Po-Yu Chen.; I-Wen Sun.; S.P. Wang. NMR evidence of hydrogen bonding in 1-ethyl-3-methylimidazolium-tetrafluoroborate room temperature ionic liquid. Inorg. Chim. Acta. 2001, 320 (1-2), 7-11.