beta-Carotene, 99%
beta-Carotene, 99%
beta-Carotene, 99%
Thermo Scientific Chemicals

beta-Carotene, 99%

CAS: 7235-40-7 | C40H56 | 536.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H60106.03
also known as H60106-03
Price (EUR)
39,65
キャンペーン価格
44,10
Save 4,45 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
39,65
キャンペーン価格
44,10
Save 4,45 (10%)
Each
Chemical Identifiers
CAS7235-40-7
IUPAC Name1,3,3-trimethyl-2-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyl-18-(2,6,6-trimethylcyclohex-1-en-1-yl)octadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl]cyclohex-1-ene
Molecular FormulaC40H56
InChI KeyOENHQHLEOONYIE-JLTXGRSLSA-N
SMILESC\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The most important of the provitamins A, β-carotene can be classified as an antioxidant due to its inhibition of radical initiated peroxidation in vitro.

Solubility
Soluble in hexane, dimethyl sulfoxide, benzene, chloroform, cyclohexane. Insoluble in water.

Notes
Air & Light Sensitive. Store in a cool place.
RUO – Research Use Only
R. Peto.; R. Doll.; J. D. Buckley.; M. B. Sporn. Can dietary beta-carotene materially reduce human cancer rates? Nature. 1981, 290201 - 208.