2-(2-Bromophenyl)ethylamine, 97%
2-(2-Bromophenyl)ethylamine, 97%
2-(2-Bromophenyl)ethylamine, 97%
2-(2-Bromophenyl)ethylamine, 97%
Thermo Scientific Chemicals

2-(2-Bromophenyl)ethylamine, 97%

CAS: 65185-58-2 | C8H10BrN | 200.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H60739.09
also known as H60739-09
Price (EUR)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS65185-58-2
IUPAC Name2-(2-bromophenyl)ethan-1-aminium
Molecular FormulaC8H11BrN
InChI KeyITRNQMJXZUWZQL-UHFFFAOYSA-O
SMILES[NH3+]CCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Artis Klapars.; Sean Parris.; Kevin W. Anderson.; Stephen L. Buchwald. Synthesis of Medium Ring Nitrogen Heterocycles via a Tandem Copper-Catalyzed C-N Bond Formation-Ring-Expansion Process. J. Am. Chem. Soc. 2004, 126 (11), 3529-3533.
  2. M. V. Mezentseva.; I. S. Nikolaeva.; E. A. Golovanova.; A. N. Fomina. Synthesis and antiviral activity of 2-anilinomethyl derivatives of 5-hydroxyindole. Pharmaceutical Chemistry Journal. 1990, 24 (10), 749-751.