(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide, 97+%
(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide, 97+%
(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide, 97+%
Thermo Scientific Chemicals

(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide, 97+%

CAS: 638132-66-8 | C18H28OP2 | 322.369 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H60834.MD
also known as H60834-MD
Price (EUR)
142,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
142,00
Each
Specifications
CAS638132-66-8
Chemical Name or Material(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide
Formula Weight322.36
IUPAC Name(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1
InChI KeyORZVYNTUPREFTI-KLHDSHLOSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only