5-(Methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals
5-(Methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals
5-(Methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-(Methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H61106.03
also known as H61106-03
1 g
Catalog number H61106.03
also known as H61106-03
Price (EUR)
244,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
244,00
Each
Chemical Identifiers
CAS20873-58-9
IUPAC Name5-(methylsulfanyl)thiophene-2-carboxylic acid
Molecular FormulaC6H6O2S2
InChI KeyFWFWUSLBIIIIEN-UHFFFAOYSA-N
SMILESCSC1=CC=C(S1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only