5'-Bromo-2'-fluoroacetophenone, 98%
5'-Bromo-2'-fluoroacetophenone, 98%
5'-Bromo-2'-fluoroacetophenone, 98%
Thermo Scientific Chemicals

5'-Bromo-2'-fluoroacetophenone, 98%

CAS: 198477-89-3 | C8H6BrFO | 217.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61282.14
also known as H61282-14
Price (EUR)
285,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
285,00
Each
Chemical Identifiers
CAS198477-89-3
IUPAC Name1-(5-bromo-2-fluorophenyl)ethan-1-one
Molecular FormulaC8H6BrFO
InChI KeyXNRQIHIOKXQSPG-UHFFFAOYSA-N
SMILESCC(=O)C1=C(F)C=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
5'-Bromo-2'-fluoroacetophenone is an intermediate used in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-Bromo-2′-fluoroacetophenone is an intermediate used in pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed.
RUO – Research Use Only