Ethyl fluoroacetate, 98%
Ethyl fluoroacetate, 98%
Ethyl fluoroacetate, 98%
Thermo Scientific Chemicals

Ethyl fluoroacetate, 98%

CAS: 459-72-3 | C4H7FO2 | 106.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H61340.09
also known as H61340-09
Price (EUR)
36,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
36,00
Each
Chemical Identifiers
CAS459-72-3
IUPAC Nameethyl 2-fluoroacetate
Molecular FormulaC4H7FO2
InChI KeyVCYZVXRKYPKDQB-UHFFFAOYSA-N
SMILESCCOC(=O)CF
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.

Solubility
Decomposes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. John T. Welch.; K. Seper.; S. Eswarakrishnan.; J. Samartino. Preparation of alpha-fluoroenolates and their use in the directed aldol reaction. J. Org. Chem. 1984, 49 (24), 4720-4721.
  2. C. J. Martin.; A. I. Schepartz.; B. F. Daubert. The Preparation of 2-Heptenal and 2-Nonenal. J. Am. Chem. Soc. 1948, 70 (7), 2601-2602.