4-Bromo-3-chlorobenzotrifluoride, 98%
4-Bromo-3-chlorobenzotrifluoride, 98%
4-Bromo-3-chlorobenzotrifluoride, 98%
Thermo Scientific Chemicals

4-Bromo-3-chlorobenzotrifluoride, 98%

CAS: 402-04-0 | C7H3BrClF3 | 259.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61978.06
also known as H61978-06
Price (EUR)
53,70
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
53,70
Each
Chemical Identifiers
CAS402-04-0
IUPAC Name1-bromo-2-chloro-4-(trifluoromethyl)benzene
Molecular FormulaC7H3BrClF3
InChI KeyRVTIHGGJJMXISV-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC(Cl)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only