N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%
N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%
N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%
Thermo Scientific Chemicals

N,N'-Di-Boc-L-histidine dicyclohexylammonium salt, 95%

CAS: 31687-58-8 | C28H48N4O6 | 536.714 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62018.03
also known as H62018-03
Price (EUR)
64,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
64,60
Each
Specifications
CAS31687-58-8
Chemical Name or MaterialN,N'-Di-Boc-L-histidine dicyclohexylammonium salt
ColorWhite
EINECS Number250-764-6
Formula Weight536.7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only