Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%
Thermo Scientific Chemicals

Methyl trans-3-aminocyclohexanecarboxylate hydrochloride, 95%

CAS: 712313-64-9 | C8H16ClNO2 | 193.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62024.03
also known as H62024-03
Price (EUR)
346,80
Online Exclusive
408,00
Save 61,20 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
346,80
Online Exclusive
408,00
Save 61,20 (15%)
Each
Add to cart
Specifications
CAS712313-64-9
Chemical Name or MaterialMethyl trans-3-aminocyclohexanecarboxylate hydrochloride
Formula Weight193.67
IUPAC Namemethyl 3-aminocyclohexane-1-carboxylate hydrochloride
InChI KeyOOFXENVWECZJBF-UHFFFAOYNA-N
View more
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trans-3-aminocyclohexanecarboxylate hydrochloride used as catalyst in pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only