trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%
trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%
trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%
Thermo Scientific Chemicals

trans-4-(Boc-amino)cyclohexaneacetaldehyde, 97%

CAS: 215790-29-7 | C13H23NO3 | 241.331 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H62373.MD
also known as H62373-MD
Price (EUR)
171,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
171,00
Each
Chemical Identifiers
CAS215790-29-7
IUPAC Nametert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
Molecular FormulaC13H23NO3
InChI KeyOPGBSEXLSWYFOR-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1CCC(CC=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormSolid
Proton NMRConforms to structure
Carbon-13 NMRConforms to structure
Assay (GC)>97%
trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexaneacetaldehyde is an intermediate for the synthesis of cariprazine (C183490), a novel antipsychotic acting on dopamine D3/D2 receptors.

Solubility
Sparingly Soluble (0.41 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only