D-Alanine ethyl ester hydrochloride, 95%
Thermo Scientific Chemicals

D-Alanine ethyl ester hydrochloride, 95%

CAS: 6331-09-5 | C8H19NO2 | 161.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H62791.14
also known as H62791-14
Price (EUR)
213,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
213,00
Each
Chemical Identifiers
CAS6331-09-5
IUPAC Nameethyl 2-aminopropanoate hydrochloridyl
Molecular FormulaC5H11ClNO2
InChI KeyCYHRNDXVTAWKMO-UHFFFAOYNA-N
SMILES[Cl].CCOC(=O)C(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
D-Alanine ethyl ester hydrochloride is used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine ethyl ester hydrochloride is used as organic intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only