Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals
Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals
Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62912.06
also known as H62912-06
5 g
Catalog number H62912.06
also known as H62912-06
Price (EUR)
378,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
378,00
Each
Chemical Identifiers
CAS113400-36-5
IUPAC Name2-benzyl 1-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate
Molecular FormulaC17H21NO5
InChI KeyTZNBTMCEMLXYEM-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1C(CCC1=O)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only