O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%
O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%
O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%
Thermo Scientific Chemicals

O-(4-Methoxybenzyl)hydroxylamine hydrochloride, 98%

CAS: 876-33-5 | C8H12ClNO2 | 189.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63312.03
also known as H63312-03
Price (EUR)
74,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
74,00
Each
Chemical Identifiers
CAS876-33-5
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(4-Methoxybenzyl)hydroxylamine hydrochloride is employed in intermediate of pharmaceutical, chemical research and analysis.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Ludovic T. Maillard; Meryem Benohoud; Philippe Durand; and Bernard Badet. A New Supported Reagent for the Parallel Synthesis of Primary and Secondary O-Alkyl Hydroxylamines through a Base-Catalyzed Mitsunobu Reaction. J. Org. Chem. 2005, 70 (16), 6303-6312.