N-Fmoc-N-methyl-L-leucine, 98%
N-Fmoc-N-methyl-L-leucine, 98%
N-Fmoc-N-methyl-L-leucine, 98%
Thermo Scientific Chemicals

N-Fmoc-N-methyl-L-leucine, 98%

CAS: 103478-62-2 | C22H25NO4 | 367.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63453.06
also known as H63453-06
Price (EUR)
315,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
315,00
Each
Chemical Identifiers
CAS103478-62-2
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-4-methylpentanoic acid
Molecular FormulaC22H25NO4
InChI KeyBUJQSIPFDWLNDC-GNLPSFAGNA-N
SMILESCC(C)C[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point113-118?C
Optical Rotation-19? to -22? (c=1 in DMF)
Assay (HPLC)≥97.5%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Jürgen Scherkenbeck; Sebastian Lüttenberg; Monika Ludwig; Karin Brücher and Andreas Kotthaus. Segment Solid-Phase Total Synthesis of the Anthelmintic Cyclooctadepsipeptides PF1022A and Emodepside. European Journal of Organic Chemistry. 2012, 8 1546-1553.