2-(Trichloroacetyl)pyrrole, 99+%
2-(Trichloroacetyl)pyrrole, 99+%
2-(Trichloroacetyl)pyrrole, 99+%
Thermo Scientific Chemicals

2-(Trichloroacetyl)pyrrole, 99+%

CAS: 35302-72-8 | C6H4Cl3NO | 212.454 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63718.14
also known as H63718-14
Price (EUR)
136,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
136,00
Each
Chemical Identifiers
CAS35302-72-8
IUPAC Name2,2,2-trichloro-1-(1H-pyrrol-2-yl)ethan-1-one
Molecular FormulaC6H4Cl3NO
InChI KeyBBFDGMDENAEMKF-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)C(=O)C1=CC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to gray to brown
FormPowder or chunks
GC/MS AnalysisConforms
Assay (GC)≥99%
Proton NMRConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only