(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%
Thermo Scientific Chemicals

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%

CAS: 252049-08-4 | C27H28N2O5 | 460.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63726.03
also known as H63726-03
Price (EUR)
132,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
132,00
Each
Specifications
CAS252049-08-4
Chemical Name or Material(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid
ColorWhite
Formula Weight474.51
IUPAC Name(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid
View more
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only