L-Glutamine tert-butyl ester hydrochloride, 98%
L-Glutamine tert-butyl ester hydrochloride, 98%
L-Glutamine tert-butyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Glutamine tert-butyl ester hydrochloride, 98%

CAS: 39741-62-3 | C9H19ClN2O3 | 238.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63868.06
also known as H63868-06
Price (EUR)
534,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
534,00
Each
Chemical Identifiers
CAS39741-62-3
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
Reactant in the synthesis of rhenium-glutamine conjugates. It is mainly used in food and pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of rhenium-glutamine conjugates. It is mainly used in food and pharma industry.

Solubility
Soluble 0.3gram in 2ml water clear solution.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
RUO – Research Use Only
Rachel Huang,; Graeme Langille,; Ravanjir K. Gill,; Cindy Mei Jin Li,; Yuji Mikata,; May Q. Wong,; Donald T. Yapp ,; Tim Storr. Synthesis, characterization, and biological studies of emissive rhenium-glutamine conjugates. Journal of Biological Inorganic Chemistry. 2013, 18(7), 831-844.