(R)-2-(Fmoc-amino)butyric acid, 98%
(R)-2-(Fmoc-amino)butyric acid, 98%
(R)-2-(Fmoc-amino)butyric acid, 98%
Thermo Scientific Chemicals

(R)-2-(Fmoc-amino)butyric acid, 98%

CAS: 170642-27-0 | C19H19NO4 | 325.364 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H63923.03
also known as H63923-03
Price (EUR)
42,80
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
42,80
Each
Chemical Identifiers
CAS170642-27-0
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Molecular FormulaC19H19NO4
InChI KeyXQIRYUNKLVPVRR-UHFFFAOYNA-N
SMILESCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation+17.0 to +21.0°
Purity≥97.5%
FormSolid
Melting Point120.0-127.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only