2,5-Dimethoxy-4-methylbenzaldehyde, 97%
2,5-Dimethoxy-4-methylbenzaldehyde, 97%
2,5-Dimethoxy-4-methylbenzaldehyde, 97%
Thermo Scientific Chemicals

2,5-Dimethoxy-4-methylbenzaldehyde, 97%

CAS: 4925-88-6 | C10H12O3 | 180.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64140.14
also known as H64140-14
Price (EUR)
797,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
797,00
Each
Chemical Identifiers
CAS4925-88-6
IUPAC Name2,5-dimethoxy-4-methylbenzaldehyde
Molecular FormulaC10H12O3
InChI KeyLRSRTWLEJBIAIT-UHFFFAOYSA-N
SMILESCOC1=CC(C)=C(OC)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
Used in the preparation of phenylamine derivative as potential psychotomimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of phenylamine derivative as potential psychotomimetics.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. David E. Nichols; Charles F. Barfknecht; John P. Long; Robert T. Standridge; Henry G. Howell; Richard A. Partyka; Donald C. Dyer. Potential psychotomimetics. 2. Rigid analogs of 2,5-dimethoxy-4-methylphenylisopropylamine (DOM, STP). J. Med. Chem. 197417(2),161-166.
  2. Shigefumi Kuwahara; Mana Saito. Enantioselective total synthesis of enokipodins A-D. Tetrahedron Letters. 200445(26), 5047-5049