5,6-Dichloropyridine-2-carboxylic acid, 98%
5,6-Dichloropyridine-2-carboxylic acid, 98%
5,6-Dichloropyridine-2-carboxylic acid, 98%
Thermo Scientific Chemicals

5,6-Dichloropyridine-2-carboxylic acid, 98%

CAS: 88912-24-7 | C6H3Cl2NO2 | 191.995 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64320.03
also known as H64320-03
Price (EUR)
111,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
111,00
Each
Chemical Identifiers
CAS88912-24-7
IUPAC Name5,6-dichloropyridine-2-carboxylic acid
Molecular FormulaC6H3Cl2NO2
InChI KeyQOJNAEPFSUYAFL-UHFFFAOYSA-N
SMILESOC(=O)C1=NC(Cl)=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only