It plays an important role as intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It plays an important role as intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Qian Zhang; Jiangshan Chen; Yanxiang Cheng; Lixiang Wang; Dongge Ma; Xiabin Jing; and Fosong Wang. Novel hole-transporting materials based on 1,4-bis(carbazolyl)benzene for organic light-emitting devices. J. Mater. Chem. 200414 (5), 895-900.
- Junqiao Ding; Qi Wang; Lei Zhao; Dongge Ma; Lixiang Wang; Xiabin Jing; and Fosong Wang. Design of star-shaped molecular architectures based on carbazole and phosphine oxide moieties: towards amorphous bipolar hosts with high triplet energy for efficient blue electrophosphorescent devices. J. Mater. Chem.201020 (37), 8126-8133.