1-Bromo-3,4,5-trichlorobenzene, 98%
1-Bromo-3,4,5-trichlorobenzene, 98%
1-Bromo-3,4,5-trichlorobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-3,4,5-trichlorobenzene, 98%

CAS: 21928-51-8 | C6H2BrCl3 | 260.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64506.06
also known as H64506-06
Price (EUR)
36,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
36,30
Each
Chemical Identifiers
CAS21928-51-8
IUPAC Name5-bromo-1,2,3-trichlorobenzene
Molecular FormulaC6H2BrCl3
InChI KeyVZUMVBQMJFFYRM-UHFFFAOYSA-N
SMILESClC1=CC(Br)=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is a multipurpose intermediate commonly used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a multipurpose intermediate commonly used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only
T. Wayne Schultz. Structure-Toxicity Relationships for Benzenes Evaluated with Tetrahymena pyriformis. Chem. Res. Toxicol. 1999, 12 (12),1262-1267 .