Methyl 4-methoxypyridine-2-carboxylate, 98%
Methyl 4-methoxypyridine-2-carboxylate, 98%
Methyl 4-methoxypyridine-2-carboxylate, 98%
Thermo Scientific Chemicals

Methyl 4-methoxypyridine-2-carboxylate, 98%

CAS: 29681-43-4 | C8H9NO3 | 167.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64519.06
also known as H64519-06
Price (EUR)
63,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
63,10
Each
Chemical Identifiers
CAS29681-43-4
IUPAC Namemethyl 4-methoxypyridine-2-carboxylate
Molecular FormulaC8H9NO3
InChI KeyOJDKENGKKYVJLY-UHFFFAOYSA-N
SMILESCOC(=O)C1=NC=CC(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only