L-Isoleucine ethyl ester hydrochloride, 98%
L-Isoleucine ethyl ester hydrochloride, 98%
L-Isoleucine ethyl ester hydrochloride, 98%
L-Isoleucine ethyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Isoleucine ethyl ester hydrochloride, 98%

CAS: 56782-52-6 | C8H18ClNO2 | 195.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64640.14
also known as H64640-14
Price (EUR)
271,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
271,00
Each
Add to cart
Chemical Identifiers
CAS56782-52-6
IUPAC Nameethyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Molecular FormulaC8H18ClNO2
InChI KeyQQGRWNMNWONMOO-LEUCUCNGSA-N
SMILESCl.CCOC(=O)[C@@H](N)[C@@H](C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only