2-Bromo-6-fluorobenzaldehyde, 98%
2-Bromo-6-fluorobenzaldehyde, 98%
2-Bromo-6-fluorobenzaldehyde, 98%
2-Bromo-6-fluorobenzaldehyde, 98%
Thermo Scientific Chemicals

2-Bromo-6-fluorobenzaldehyde, 98%

CAS: 360575-28-6 | C7H4BrFO | 203.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H64689.22
also known as H64689-22
Price (EUR)
682,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
682,00
Each
Add to cart
Chemical Identifiers
CAS360575-28-6
IUPAC Name2-bromo-6-fluorobenzaldehyde
Molecular FormulaC7H4BrFO
InChI KeyPJNILWKRAKKEQM-UHFFFAOYSA-N
SMILESFC1=C(C=O)C(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Christian Ruzié; Jolanta Karpinska; Alan R. Kennedy; and Yves H. Geerts. Synthesis of 1,6-, 2,7-, 3,8-, and 4,9-Isomers of Didodecyl[1]benzothieno[3,2-b][1]benzothiophenes. J. Org. Chem. 2013 78(15), 7741-7748.
  2. Xinzhi Chen; Shaodong Zhou; Chao Qian; Chaohong He. Synthesis of 1H-indazole: a combination of experimental and theoretical studies. Research on Chemical Intermediates. 2013 38(7), 1619-1628.