Ethyl 2-chloropyrimidine-5-carboxylate, 97%
Ethyl 2-chloropyrimidine-5-carboxylate, 97%
Ethyl 2-chloropyrimidine-5-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 2-chloropyrimidine-5-carboxylate, 97%

CAS: 89793-12-4 | C7H7ClN2O2 | 186.595 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64914.03
also known as H64914-03
Price (EUR)
70,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
70,00
Each
Chemical Identifiers
CAS89793-12-4
IUPAC Nameethyl 2-chloropyrimidine-5-carboxylate
Molecular FormulaC7H7ClN2O2
InChI KeyIEMKQRSOAOPKRJ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CN=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC/MS AnalysisConforms
Appearance (Color)Pale cream to yellow
Assay (GC)≥96%
Identification (FTIR)Conforms
Proton NMRConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only