3-Cyanobenzeneboronic acid pinacol ester, 97%
3-Cyanobenzeneboronic acid pinacol ester, 97%
3-Cyanobenzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-Cyanobenzeneboronic acid pinacol ester, 97%

CAS: 214360-46-0 | C13H16BNO2 | 229.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64948.03
also known as H64948-03
Price (EUR)
43,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
43,30
Each
Chemical Identifiers
CAS214360-46-0
IUPAC Name3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular FormulaC13H16BNO2
InChI KeyFIGQEPXOSAFKTA-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only