2-Amino-5-chlorophenol, 97%
2-Amino-5-chlorophenol, 97%
2-Amino-5-chlorophenol, 97%
Thermo Scientific Chemicals

2-Amino-5-chlorophenol, 97%

CAS: 28443-50-7 | C6H6ClNO | 143.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H64978.14
also known as H64978-14
Price (EUR)
85,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
85,00
Each
Chemical Identifiers
CAS28443-50-7
IUPAC Name2-amino-5-chlorophenol
Molecular FormulaC6H6ClNO
InChI KeyFZCQMIRJCGWWCL-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay from Supplier's CofA≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only