PTQ10
Thermo Scientific Chemicals

PTQ10

CAS: 2270233-86-6 | (C{2}{8}H{3}{6}F{2}N{2}OS)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
500 mg
Catalog number H66106.MC
also known as H66106-MC
Price (EUR)
517,00
Each
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
517,00
Each
Chemical Identifiers
CAS2270233-86-6
SpecificationsSpecification SheetSpecification Sheet
FormFlake or threadlike
Appearance (Color)Black
Bandgap energy1.96 ? 0.02 eV
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only