3-Chloro-N-Fmoc-L-phenylalanine, 95%
3-Chloro-N-Fmoc-L-phenylalanine, 95%
3-Chloro-N-Fmoc-L-phenylalanine, 95%
Thermo Scientific Chemicals

3-Chloro-N-Fmoc-L-phenylalanine, 95%

CAS: 198560-44-0 | C24H20ClNO4 | 421.877 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66237.03
also known as H66237-03
Price (EUR)
57,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
57,10
Each
Specifications
CAS198560-44-0
Chemical Name or Material3-Chloro-N-Fmoc-L-phenylalanine
Formula Weight421.88
IUPAC Name(2S)-3-(3-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
InChI KeyUOZAKKJRIKXQPY-QFIPXVFZSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only