2-Amino-3-chlorobenzoic acid, 98%
2-Amino-3-chlorobenzoic acid, 98%
2-Amino-3-chlorobenzoic acid, 98%
Thermo Scientific Chemicals

2-Amino-3-chlorobenzoic acid, 98%

CAS: 6388-47-2 | C7H6ClNO2 | 171.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H66261.14
also known as H66261-14
Price (EUR)
76,20
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
76,20
Each
Chemical Identifiers
CAS6388-47-2
IUPAC Name2-amino-3-chlorobenzoic acid
Molecular FormulaC7H6ClNO2
InChI KeyLWUAMROXVQLJKA-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
2-Amino-3-chlorobenzoic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-3-chlorobenzoic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Air & Light Sensitive. Store away from air. Store in the dark. Incompatible with light, air and oxidizing agents.
RUO – Research Use Only
Jungkul Lee; Michael Simurdiak and Huimin Zhao. Reconstitution and Characterization of Aminopyrrolnitrin Oxygenase, a Rieske N-Oxygenase That Catalyzes Unusual Arylamine Oxidation. Journal of Biological Chemistry.2005, 280, 36719-36727.