(S)-(-)-Tetrahydro-3-furoic acid, 97%
(S)-(-)-Tetrahydro-3-furoic acid, 97%
(S)-(-)-Tetrahydro-3-furoic acid, 97%
Thermo Scientific Chemicals

(S)-(-)-Tetrahydro-3-furoic acid, 97%

CAS: 168395-26-4 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H66392.MD
also known as H66392-MD
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS168395-26-4
IUPAC Nameoxolane-3-carboxylic acid
Molecular FormulaC5H8O3
InChI KeyBOTREHHXSQGWTR-UHFFFAOYNA-N
SMILESOC(=O)C1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
Assay (GC)≥96.0%
Assay from Supplier's CofASpecific optical rotation [α]= -25.76? c=0.747g/ 100 mL acetone
GC/MS AnalysisConforms
Proton NMRConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only