N-Boc-D-aspartic acid 1-methyl ester, 97%
N-Boc-D-aspartic acid 1-methyl ester, 97%
N-Boc-D-aspartic acid 1-methyl ester, 97%
Thermo Scientific Chemicals

N-Boc-D-aspartic acid 1-methyl ester, 97%

CAS: 137130-65-5 | C10H17NO6 | 247.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66573.06
also known as H66573-06
Price (EUR)
83,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
83,90
Each
Chemical Identifiers
CAS137130-65-5
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (HPLC)>96.0%
N-Boc-D-aspartic acid 1-methyl ester is widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-D-aspartic acid 1-methyl ester is widely used in pharmaceuticals and foods.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only