Tetramethyl orthocarbonate, 95%
Tetramethyl orthocarbonate, 95%
Tetramethyl orthocarbonate, 95%
Thermo Scientific Chemicals

Tetramethyl orthocarbonate, 95%

CAS: 1850-14-2 | C5H12O4 | 136.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66617.03
also known as H66617-03
Price (EUR)
46,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
46,10
Each
Chemical Identifiers
CAS1850-14-2
IUPAC Nametetramethoxymethane
Molecular FormulaC5H12O4
InChI KeyAHJWSRRHTXRLAQ-UHFFFAOYSA-N
SMILESCOC(OC)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)96.1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only