1-Ethylcyclopentanol, 96%, Thermo Scientific Chemicals
1-Ethylcyclopentanol, 96%, Thermo Scientific Chemicals
1-Ethylcyclopentanol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Ethylcyclopentanol, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H66809.06
also known as H66809-06
5 g
Catalog number H66809.06
also known as H66809-06
Price (EUR)
100,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
100,00
Each
Specifications
CAS1462-96-0
Chemical Name or Material1-Ethylcyclopentanol
ColorColorless to Yellow
DOT InformationTransport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Density0.909 g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only