4-Nitrophenyl-alpha-L-Fucopyranoside
4-Nitrophenyl-alpha-L-Fucopyranoside
4-Nitrophenyl-alpha-L-Fucopyranoside
Thermo Scientific Chemicals

4-Nitrophenyl-alpha-L-Fucopyranoside

CAS: 10231-84-2285.252 g/mol
Have Questions?
Catalog NumberQuantity
J34131.031 g
Catalog number J34131.03
Price (EUR)
541,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
541,00
Each
Chemical Identifiers
CAS10231-84-2
IUPAC Name(2S,3S,4R,5S,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular FormulaC12H15NO7
InChI KeyYILIDCGSXCGACV-SQKFTNEHSA-N
SMILESC[C@@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@@H](O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormWhite powder
Melting Point196? ? 2?C
 
RUO – Research Use Only