Separating or resolving buffer (4X)
Thermo Scientific Chemicals

Separating or resolving buffer (4X)

Have Questions?
Change viewbuttonViewtableView
Quantity:
500mL
1L
Catalog number J60181.K2
also known as J60181-K2
Price (EUR)
90,65
線上優惠
101,00
Save 10,35 (10%)
Each
Quantity:
1L
Request bulk or custom format
Price (EUR)
90,65
線上優惠
101,00
Save 10,35 (10%)
Each
Specifications
Concentration4X
FormLiquid
Quantity1L
Storage RequirementsAmbient temperatures
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only