Yohimbine hydrochloride, 98+%
Yohimbine hydrochloride, 98+%
Yohimbine hydrochloride, 98+%
Thermo Scientific Chemicals

Yohimbine hydrochloride, 98+%

CAS: 65-19-0 | C21H27ClN2O3, C21H26N2O3·HCl | 390.91 g/mol
Have Questions?
Catalog NumberQuantity
J60185.03
also known as J60185-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60185.03
also known as J60185-03
Price (EUR)
62,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
62,70
Each
Chemical Identifiers
CAS65-19-0
IUPAC Namehydrogen methyl (1S,15R,18S,19R,20S)-18-hydroxy-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate chloride
Molecular FormulaC21H27ClN2O3
InChI KeyPIPZGJSEDRMUAW-VJDCAHTMSA-N
SMILES[H+].[Cl-].COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0%
FormPowder
Yohimbine hydrochloride is an α2-adrenergic receptor antagonist. It blocks the central α2-adrenegic receptors in the brain, thus preventing and reducing the effects of xylazine, an α2-adrenergic agonist. The sedative effects and respiratory depression that accompany xylazine administration are both antagonized and reversed by yohimbine. Yohimbine hydrochloride also produces an antidiuretic effect, and increases heart rate and blood pressure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only