U0126, 99+%
U0126, 99+%
U0126, 99+%
Thermo Scientific Chemicals

U0126, 99+%

Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2 | CAS: 109511-58-2 | C18H16N6S2, C18H16N6S2 | 380.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J61246.MB
also known as J61246-MB
Price (EUR)
144,65
キャンペーン価格
160,00
Save 15,35 (10%)
Each
Quantity:
25 mg
Request bulk or custom format
Price (EUR)
144,65
キャンペーン価格
160,00
Save 15,35 (10%)
Each
Chemical Identifiers
CAS109511-58-2
IUPAC Name(2Z,3Z)-bis({amino[(2-aminophenyl)sulfanyl]methylidene})butanedinitrile
Molecular FormulaC18H16N6S2
InChI KeyDVEXZJFMOKTQEZ-JYFOCSDGSA-N
SMILESN\C(SC1=CC=CC=C1N)=C(\C#N)/C(/C#N)=C(\N)SC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.0% (TLC)
Appearance (Color)White to off-white
Assay from Supplier's CofA≥99.0% (HPLC)
FormPowder or crystals or crystalline powder or granules
Melting Point130.0-133.0°C
U0126, is used as a selective MAP Kinase Kinase (MKK, MEK) inhibitor. U-0126 has been observed to inhibit promoters containing an AP-1 response element, while in contrast having no effect on promoters that lack an AP-1 response group. U-0126 is also useful in neuronal studies in mice since it displays inhibitory effects against oxidative stress. U-0126 is an activator of PGC-1, mtTFA and nuclear respiratory factor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C

Frequently asked questions (FAQs)

What is U0126, ≥99% (Cat. No. J61246.MC, J61246.MB) soluble in?

U0126, ≥99% (Cat. No. J61246.MC, J61246.MB) is soluble in DMSO. It is poorly soluble in ethanol and water.