(+)-1-Deoxymannojirimycin hydrochloride, Thermo Scientific Chemicals
(+)-1-Deoxymannojirimycin hydrochloride, Thermo Scientific Chemicals
(+)-1-Deoxymannojirimycin hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+)-1-Deoxymannojirimycin hydrochloride, Thermo Scientific Chemicals

Selective inhibitor of a-mannosidase
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J61277.LB0
also known as J61277-#0
Price (EUR)
166,00
Each
Quantity:
5 mg
Request bulk or custom format
Price (EUR)
166,00
Each
Specifications
CAS73465-43-7
Chemical Name or Material(+)-1-Deoxymannojirimycin hydrochloride
Formula Weight199.6
IUPAC Name(2R,3R,4R,5R)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
InChI KeyZJIHMALTJRDNQI-MVNLRXSJSA-N
View more
Selective inhibitor of a-mannosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only